Anti-DRD1 Mouse Monoclonal Antibody [clone: 887438]

Supplier: R & D Systems
MAB8276
102887-556EA 594.37 USD
102887-556
Anti-DRD1 Mouse Monoclonal Antibody [clone: 887438]
Antibodies
Type: Primary
Antigen: DRD1
Clonality: Monoclonal
Clone: 887438
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR