Anti-BCL2 Mouse Monoclonal Antibody [clone: 118712]

Supplier: R & D Systems
MAB8271
220021-586EA 608.83 USD
220021-586
Anti-BCL2 Mouse Monoclonal Antibody [clone: 118712]
Antibodies
Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 118712
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR