Anti-BCL2 Mouse Monoclonal Antibody [clone: 118701]

Supplier: R & D Systems
MAB827
220021-585EA 639.2 USD
220021-585
Anti-BCL2 Mouse Monoclonal Antibody [clone: 118701]
Antibodies
Simple Western

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 118701
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR