Anti-B2M Mouse Monoclonal Antibody [clone: 883028]

Supplier: R & D Systems
MAB8248
102887-550EA 623.29 USD
102887-550
Anti-B2M Mouse Monoclonal Antibody [clone: 883028]
Antibodies
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: 883028
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR