Anti-MUC19 Mouse Monoclonal Antibody [clone: 876013]

Supplier: R & D Systems
MAB8245
102887-548EA 594.37 USD
102887-548
Anti-MUC19 Mouse Monoclonal Antibody [clone: 876013]
Antibodies
Type: Primary
Antigen: MUC19
Clonality: Monoclonal
Clone: 876013
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR