Anti-BCL2L2 Mouse Monoclonal Antibody [clone: 90336]

Supplier: R & D Systems
MAB8241
220021-584EA 584.25 USD
220021-584
Anti-BCL2L2 Mouse Monoclonal Antibody [clone: 90336]
Antibodies
Type: Primary
Antigen: BCL2L2
Clonality: Monoclonal
Clone: 90336
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR