Anti-LGR5 Rat Monoclonal Antibody [clone: 889901]

Supplier: R & D Systems
MAB82401
102887-546EA 594.37 USD
102887-546
Anti-LGR5 Rat Monoclonal Antibody [clone: 889901]
Antibodies
Type: Primary
Antigen: LGR5
Clonality: Monoclonal
Clone: 889901
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR