Anti-LGR5 Rat Monoclonal Antibody [clone: 803420]

Supplier: R & D Systems
MAB8240
102887-544EA 633.42 USD
102887-544
Anti-LGR5 Rat Monoclonal Antibody [clone: 803420]
Antibodies
Agonist Activity

Type: Primary
Antigen: LGR5
Clonality: Monoclonal
Clone: 803420
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR