Anti-SLC14A1 Mouse Monoclonal Antibody [clone: 888418]

Supplier: R & D Systems
MAB8238
102887-542EA 577.02 USD
102887-542
Anti-SLC14A1 Mouse Monoclonal Antibody [clone: 888418]
Antibodies
Type: Primary
Antigen: SLC14A1
Clonality: Monoclonal
Clone: 888418
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR