Anti-SERPINA11 Mouse Monoclonal Antibody [clone: 895842]

Supplier: R & D Systems
MAB8236
102887-540EA 561.11 USD
102887-540
Anti-SERPINA11 Mouse Monoclonal Antibody [clone: 895842]
Antibodies
Type: Primary
Antigen: SERPINA11
Clonality: Monoclonal
Clone: 895842
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR