Anti-CASP7 Mouse Monoclonal Antibody [clone: MCH3101.62]

Supplier: R & D Systems
MAB823
220021-582EA 642.09 USD
220021-582
Anti-CASP7 Mouse Monoclonal Antibody [clone: MCH3101.62]
Antibodies
Type: Primary
Antigen: CASP7
Clonality: Monoclonal
Clone: MCH3101.62
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR