Anti-MAF Mouse Monoclonal Antibody [clone: 883926]

Supplier: R & D Systems
MAB8227
102887-538EA 594.37 USD
102887-538
Anti-MAF Mouse Monoclonal Antibody [clone: 883926]
Antibodies
Type: Primary
Antigen: MAF
Clonality: Monoclonal
Clone: 883926
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR