Anti-XIAP Mouse Monoclonal Antibody [clone: 117318]

Supplier: R & D Systems
MAB8221
220021-581EA 577.02 USD
220021-581
Anti-XIAP Mouse Monoclonal Antibody [clone: 117318]
Antibodies
Type: Primary
Antigen: XIAP
Clonality: Monoclonal
Clone: 117318
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR