Anti-ANGPT1 Rat Monoclonal Antibody [clone: 892732]

Supplier: R & D Systems
MAB8220
102887-534EA 633.42 USD
102887-534
Anti-ANGPT1 Rat Monoclonal Antibody [clone: 892732]
Antibodies
Type: Primary
Antigen: ANGPT1
Clonality: Monoclonal
Clone: 892732
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR