Anti-XIAP Mouse Monoclonal Antibody [clone: 117320]

Supplier: R & D Systems
MAB822
220021-580EA 594.37 USD
220021-580
Anti-XIAP Mouse Monoclonal Antibody [clone: 117320]
Antibodies
Type: Primary
Antigen: XIAP
Clonality: Monoclonal
Clone: 117320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR