Anti-CASP14 Mouse Monoclonal Antibody [clone: 868715]

Supplier: R & D Systems
MAB8215
102887-532EA 594.37 USD
102887-532
Anti-CASP14 Mouse Monoclonal Antibody [clone: 868715]
Antibodies
Simple Western

Type: Primary
Antigen: CASP14
Clonality: Monoclonal
Clone: 868715
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR