Anti-GIPR Mouse Monoclonal Antibody [clone: 591853]

Supplier: R & D Systems
MAB8210
102887-530EA 485.91 USD
102887-530
Anti-GIPR Mouse Monoclonal Antibody [clone: 591853]
Antibodies
Type: Primary
Antigen Symbol: GIPR
Antigen Name: gastric inhibitory polypeptide receptor
Antigen Synonyms: PGQTL2
Clonality: Monoclonal
Clone: 591853
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR