Anti-KPNB1 Mouse Monoclonal Antibody [clone: 845208]

Supplier: R & D Systems
MAB8209
102887-528EA 594.37 USD
102887-528
Anti-KPNB1 Mouse Monoclonal Antibody [clone: 845208]
Antibodies
Type: Primary
Antigen: KPNB1
Clonality: Monoclonal
Clone: 845208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR