Anti-MC5R Mouse Monoclonal Antibody [clone: 821012]

Supplier: R & D Systems
MAB8205
102887-526EA 577.02 USD
102887-526
Anti-MC5R Mouse Monoclonal Antibody [clone: 821012]
Antibodies
Type: Primary
Antigen: MC5R
Clonality: Monoclonal
Clone: 821012
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR