Anti-KPNA4 Mouse Monoclonal Antibody [clone: 843124]

Supplier: R & D Systems
MAB8204
102887-524EA 594.37 USD
102887-524
Anti-KPNA4 Mouse Monoclonal Antibody [clone: 843124]
Antibodies
Simple Western

Type: Primary
Antigen: KPNA4
Clonality: Monoclonal
Clone: 843124
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR