Anti-GHRL Rat Monoclonal Antibody [clone: 883622]

Supplier: R & D Systems
MAB8200
102887-522EA 594.37 USD
102887-522
Anti-GHRL Rat Monoclonal Antibody [clone: 883622]
Antibodies
Type: Primary
Antigen Symbol: GHRL
Antigen Name: ghrelin
Antigen Synonyms: 2210006E23Rik, Ghr, MTLRP, MTLRPAP, m46
Clonality: Monoclonal
Clone: 883622
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR