Anti-MUC4 Mouse Monoclonal Antibody [clone: 781631]

Supplier: R & D Systems
MAB8195
102887-518EA 577.02 USD
102887-518
Anti-MUC4 Mouse Monoclonal Antibody [clone: 781631]
Antibodies
Type: Primary
Antigen: MUC4
Clonality: Monoclonal
Clone: 781631
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR