Anti-BIRC2 Mouse Monoclonal Antibody [clone: 681724]

Supplier: R & D Systems
MAB8181
102887-516EA 594.37 USD
102887-516
Anti-BIRC2 Mouse Monoclonal Antibody [clone: 681724]
Antibodies
Type: Primary
Antigen: BIRC2
Clonality: Monoclonal
Clone: 681724
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR