Anti-SERPINA7 Mouse Monoclonal Antibody [clone: 893339]

Supplier: R & D Systems
MAB8176
102887-512EA 594.37 USD
102887-512
Anti-SERPINA7 Mouse Monoclonal Antibody [clone: 893339]
Antibodies
Type: Primary
Antigen: SERPINA7
Clonality: Monoclonal
Clone: 893339
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR