Anti-MFI2 Mouse Monoclonal Antibody [clone: 893416]

Supplier: R & D Systems
MAB81751
102887-510EA 600.15 USD
102887-510
Anti-MFI2 Mouse Monoclonal Antibody [clone: 893416]
Antibodies
Type: Primary
Antigen: MFI2
Clonality: Monoclonal
Clone: 893416
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR