Anti-GSN Mouse Monoclonal Antibody [clone: 893205]

Supplier: R & D Systems
MAB8170
102887-506EA 594.37 USD
102887-506
Anti-GSN Mouse Monoclonal Antibody [clone: 893205]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: GSN
Antigen Name: gelsolin
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 893205
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR