Anti-BIRC3 Mouse Monoclonal Antibody [clone: 315304]

Supplier: R & D Systems
MAB817
220021-578EA 639.2 USD
220021-578
Anti-BIRC3 Mouse Monoclonal Antibody [clone: 315304]
Antibodies
Simple Western

Type: Primary
Antigen: BIRC3
Clonality: Monoclonal
Clone: 315304
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR