Anti-MAPKAP1 Mouse Monoclonal Antibody [clone: 752219]

Supplier: R & D Systems
MAB8168
102887-502EA 561.11 USD
102887-502
Anti-MAPKAP1 Mouse Monoclonal Antibody [clone: 752219]
Antibodies
Type: Primary
Antigen: MAPKAP1
Clonality: Monoclonal
Clone: 752219
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR