Anti-SEMA4A Rat Monoclonal Antibody [clone: 757129]

Supplier: R & D Systems
MAB8166
102887-498EA 594.37 USD
102887-498
Anti-SEMA4A Rat Monoclonal Antibody [clone: 757129]
Antibodies
Type: Primary
Antigen: SEMA4A
Clonality: Monoclonal
Clone: 757129
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR