Anti-TM4SF1 Mouse Monoclonal Antibody [clone: 877621]

Supplier: R & D Systems
MAB8164
102887-496EA 623.29 USD
102887-496
Anti-TM4SF1 Mouse Monoclonal Antibody [clone: 877621]
Antibodies
Type: Primary
Antigen: TM4SF1
Clonality: Monoclonal
Clone: 877621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR