Anti-BAK Mouse Monoclonal Antibody [clone: 564305]

Supplier: R & D Systems
MAB8161
220021-577EA 594.37 USD
220021-577
Anti-BAK Mouse Monoclonal Antibody [clone: 564305]
Antibodies
Type: Primary
Antigen: BAK
Clonality: Monoclonal
Clone: 564305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR