Anti-HCN4 Mouse Monoclonal Antibody [clone: 880904]

Supplier: R & D Systems
MAB8138
102887-494EA 594.37 USD
102887-494
Anti-HCN4 Mouse Monoclonal Antibody [clone: 880904]
Antibodies
Type: Primary
Antigen Symbol: HCN4
Antigen Name: hyperpolarization activated cyclic nucleotide gated potassium channel 4
Antigen Synonyms: SSS2
Clonality: Monoclonal
Clone: 880904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR