Anti-CASP8 Mouse Monoclonal Antibody [clone: 746109]

Supplier: R & D Systems
MAB8135
102887-490EA 584.25 USD
102887-490
Anti-CASP8 Mouse Monoclonal Antibody [clone: 746109]
Antibodies
Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone: 746109
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR