Anti-EGFR Rat Monoclonal Antibody [clone: 869111]

Supplier: R & D Systems
MAB8130
102887-488EA 577.02 USD
102887-488
Anti-EGFR Rat Monoclonal Antibody [clone: 869111]
Antibodies
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 869111
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR