Anti-ICAM3 Mouse Monoclonal Antibody [clone: 76205]

Supplier: R & D Systems
MAB813
220021-575EA 772.25 USD
220021-575
Anti-ICAM3 Mouse Monoclonal Antibody [clone: 76205]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: 76205
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR