Anti-C9 Mouse Monoclonal Antibody [clone: 887025]

Supplier: R & D Systems
MAB8126
102887-486EA 582.8 USD
102887-486
Anti-C9 Mouse Monoclonal Antibody [clone: 887025]
Antibodies
Simple Western

Type: Primary
Antigen: C9
Clonality: Monoclonal
Clone: 887025
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR