Anti-TBX20 Mouse Monoclonal Antibody [clone: 668710]

Supplier: R & D Systems
MAB8124
102887-484EA 594.37 USD
102887-484
Anti-TBX20 Mouse Monoclonal Antibody [clone: 668710]
Antibodies
Type: Primary
Antigen: TBX20
Clonality: Monoclonal
Clone: 668710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR