Anti-CXCR2 Mouse Monoclonal Antibody [clone: 866614]

Supplier: R & D Systems
MAB8110
102887-480EA 561.11 USD
102887-480
Anti-CXCR2 Mouse Monoclonal Antibody [clone: 866614]
Antibodies
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 866614
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR