Anti-SHARPIN Mouse Monoclonal Antibody [clone: 881816]

Supplier: R & D Systems
MAB8100
102887-476EA 584.25 USD
102887-476
Anti-SHARPIN Mouse Monoclonal Antibody [clone: 881816]
Antibodies
Type: Primary
Antigen: SHARPIN
Clonality: Monoclonal
Clone: 881816
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR