Anti-BCL2 Rat Monoclonal Antibody [clone: 121529]

Supplier: R & D Systems
MAB810
220021-571EA 594.37 USD
220021-571
Anti-BCL2 Rat Monoclonal Antibody [clone: 121529]
Antibodies
Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 121529
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR