Anti-MIXL1 Rat Monoclonal Antibody [clone: 868132]

Supplier: R & D Systems
MAB8099
102887-474EA 577.02 USD
102887-474
Anti-MIXL1 Rat Monoclonal Antibody [clone: 868132]
Antibodies
Type: Primary
Antigen: MIXL1
Clonality: Monoclonal
Clone: 868132
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR