Anti-CBX2 Mouse Monoclonal Antibody [clone: 880833]

Supplier: R & D Systems
MAB8098
102887-472EA 561.11 USD
102887-472
Anti-CBX2 Mouse Monoclonal Antibody [clone: 880833]
Antibodies
Type: Primary
Antigen: CBX2
Clonality: Monoclonal
Clone: 880833
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR