Anti-PRDM14 Rat Monoclonal Antibody [clone: 856235]

Supplier: R & D Systems
MAB8097
102887-470EA 594.37 USD
102887-470
Anti-PRDM14 Rat Monoclonal Antibody [clone: 856235]
Antibodies
Type: Primary
Antigen: PRDM14
Clonality: Monoclonal
Clone: 856235
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR