Anti-PTCH2 Rat Monoclonal Antibody [clone: 865619]

Supplier: R & D Systems
MAB8096
102887-468EA 577.02 USD
102887-468
Anti-PTCH2 Rat Monoclonal Antibody [clone: 865619]
Antibodies
Type: Primary
Antigen: PTCH2
Clonality: Monoclonal
Clone: 865619
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR