Anti-PARP1 Mouse Monoclonal Antibody [clone: 839120]

Supplier: R & D Systems
MAB8095
102887-466EA 639.2 USD
102887-466
Anti-PARP1 Mouse Monoclonal Antibody [clone: 839120]
Antibodies
Simple Western

Type: Primary
Antigen: PARP1
Clonality: Monoclonal
Clone: 839120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR