Anti-YAP1 Mouse Monoclonal Antibody [clone: 867711]

Supplier: R & D Systems
MAB8094
102887-464EA 594.37 USD
102887-464
Anti-YAP1 Mouse Monoclonal Antibody [clone: 867711]
Antibodies
Simple Western

Type: Primary
Antigen: YAP1
Clonality: Monoclonal
Clone: 867711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR