Anti-LGR5 Mouse Monoclonal Antibody [clone: 707042]

Supplier: R & D Systems
MAB8078
102887-462EA 655.11 USD
102887-462
Anti-LGR5 Mouse Monoclonal Antibody [clone: 707042]
Antibodies
Agonist Activity

Type: Primary
Antigen: LGR5
Clonality: Monoclonal
Clone: 707042
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR