Anti-PREX1 Mouse Monoclonal Antibody [clone: 869704]

Supplier: R & D Systems
MAB8076
102887-460EA 577.02 USD
102887-460
Anti-PREX1 Mouse Monoclonal Antibody [clone: 869704]
Antibodies
Simple Western

Type: Primary
Antigen: PREX1
Clonality: Monoclonal
Clone: 869704
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR