Anti-SATB1 Mouse Monoclonal Antibody [clone: 815427]

Supplier: R & D Systems
MAB8075
102887-458EA 594.37 USD
102887-458
Anti-SATB1 Mouse Monoclonal Antibody [clone: 815427]
Antibodies
Simple Western

Type: Primary
Antigen: SATB1
Clonality: Monoclonal
Clone: 815427
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR