Anti-ITLN1 Rat Monoclonal Antibody [clone: 746420]

Supplier: R & D Systems
MAB8074
102887-456EA 594.37 USD
102887-456
Anti-ITLN1 Rat Monoclonal Antibody [clone: 746420]
Antibodies
Type: Primary
Antigen: ITLN1
Clonality: Monoclonal
Clone: 746420
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR